Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038704.2 | NP_001033793.2 | 1 | 247 | ||
XM_011524637.2 | XP_011522939.1 | 1 | 247 | |||
XM_017024498.1 | XP_016879987.1 | 1 | 247 | |||
GenBank mRNA | AK097440.1 | - | 1 | 197 | ||
BC022189.2 | - | 1 | 243 | 4824904 | ||
DB073141.1 | - | 1 | 249 |