Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173625.4 | NP_775896.3 | 4 | 515 | ||
XM_011524647.2 | XP_011522949.1 | 3 | 704 | |||
XM_011524649.2 | XP_011522951.1 | 4 | 789 | |||
GenBank mRNA | BC034672.1 | - | 4 | 458 | 5184947 | |
EG328593.1 | - | 3 | 318 |