Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17orf78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321399.1 | NP_001308328.1 | 3 | 282 | ||
NM_173625.4 | NP_775896.3 | 3 | 282 | |||
XM_011524647.2 | XP_011522949.1 | 2 | 471 | |||
XM_011524648.2 | XP_011522950.1 | 3 | 556 | |||
XM_011524649.2 | XP_011522951.1 | 3 | 556 | |||
GenBank mRNA | AK096966.1 | - | 3 | 283 | ||
BC034672.1 | - | 3 | 225 | 5184947 |