Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178171.4 | NP_835465.2 | 10, 11 | 1139 | ||
XM_006721832.2 | XP_006721895.1 | 10, 11 | 1092 | |||
XM_011524651.2 | XP_011522953.1 | 8, 9 | 855 | |||
XM_017024502.1 | XP_016879991.1 | 9, 10 | 1112 | |||
GenBank mRNA | AB093591.1 | - | 9, 10 | 1032 | ||
AF307953.1 | - | 9, 10 | 1054 | |||
AK096439.1 | - | 11, 12 | 1141 | |||
BC109197.2 | - | 11, 12 | 1101 | 40005562 |