Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSDMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178171.4 | NP_835465.2 | 3 | 413 | ||
XM_006721832.2 | XP_006721895.1 | 3 | 366 | |||
XM_017024502.1 | XP_016879991.1 | 3 | 413 | |||
XM_017024503.1 | XP_016879992.1 | 3 | 413 | |||
XM_017024504.1 | XP_016879993.1 | 3 | 413 | |||
GenBank mRNA | AB093591.1 | - | 2 | 306 | ||
AF307953.1 | - | 2 | 325 | |||
AK096439.1 | - | 3 | 415 | |||
BC109197.2 | - | 3 | 375 | 40005562 |