Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: F11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000128.3 | NP_000119.1 | 6 | 894 | ||
XM_005262821.3 | XP_005262878.1 | 6 | 913 | |||
XM_005262822.3 | XP_005262879.1 | 6 | 913 | |||
XM_006714137.2 | XP_006714200.1 | 6 | 913 | |||
XM_017007884.1 | XP_016863373.1 | 6 | 913 | |||
XM_017007885.1 | XP_016863374.1 | 6 | 913 | |||
XM_017007886.1 | XP_016863375.1 | 6 | 913 | |||
GenBank mRNA | AF045649.1 | - | 5 | 433 | ||
AK313195.1 | - | 6 | 665 | |||
BC119014.1 | - | 6 | 657 | 40085167 | ||
BC122863.1 | - | 6 | 657 | 40085169 | ||
M13142.1 | - | 6 | 604 |