Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC26A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166347.1 | NP_001159819.1 | 13 | 1503 | ||
NM_001166348.1 | NP_001159820.1 | 12 | 1433 | |||
NM_001166349.1 | NP_001159821.1 | 12 | 1303 | |||
NM_173626.3 | NP_775897.3 | 13 | 1500 | |||
XM_006721833.1 | XP_006721896.1 | 13 | 1575 | |||
XM_011524652.1 | XP_011522954.1 | 11 | 1336 | |||
XM_011524653.1 | XP_011522955.1 | 11 | 1264 | |||
XM_011524654.1 | XP_011522956.1 | 11 | 1149 | |||
XM_017024505.1 | XP_016879994.1 | 12 | 1489 | |||
XM_017024506.1 | XP_016879995.1 | 12 | 1489 | |||
XM_017024507.1 | XP_016879996.1 | 13 | 1549 | |||
GenBank mRNA | AF331524.1 | - | 5 | 277 | ||
AF345195.1 | - | 12 | 1235 | |||
AJ544073.1 | - | 13 | 1474 | |||
AK075248.1 | - | 13 | 1463 | |||
AK315132.1 | - | 13 | 1466 | |||
AL833468.1 | - | 12 | 1397 | |||
BC035900.1 | - | 13 | 1442 | 5241643 | ||
BC047451.1 | - | 7 | 552 |