Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182572.3 | NP_872378.3 | 4 | 663 | ||
XM_006723149.2 | XP_006723212.1 | 3 | 2276 | |||
XM_017026609.1 | XP_016882098.1 | 4 | 1222 | |||
XM_017026610.1 | XP_016882099.1 | 4 | 663 | |||
GenBank mRNA | AK091098.1 | - | 4 | 663 |