Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127505.2 | NP_001120977.1 | 14 | 1302 | ||
NM_004315.5 | NP_004306.3 | 14 | 1368 | |||
NM_177924.4 | NP_808592.2 | 14 | 1435 | |||
XM_005273504.3 | XP_005273561.1 | 14 | 1196 | |||
GenBank mRNA | AK025732.1 | - | 15 | 1338 | ||
AK098299.1 | - | 14 | 1083 | |||
AK222780.1 | - | 15 | 1171 | |||
AK289481.1 | - | 15 | 1336 | |||
AY305384.1 | - | 14 | 1209 | |||
BC016481.1 | - | 15 | 1158 | 3923451 | ||
BC016828.1 | - | 14 | 1289 | 4109317 | ||
BC035453.1 | - | 9 | 733 | |||
U47674.1 | - | 14 | 1139 | |||
U70063.1 | - | 15 | 1140 |