Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321864.1 | NP_001308793.1 | 8, 9 | 887 | ||
NM_001321865.1 | NP_001308794.1 | 8, 9 | 1050 | |||
NM_182575.2 | NP_872381.2 | 9, 10 | 1488 | |||
NR_135832.1 | 8, 9 | 976 | ||||
XM_005258793.4 | XP_005258850.1 | 9, 10 | 1221 | |||
XM_011526811.2 | XP_011525113.1 | 7, 8 | 1911 | |||
XM_011526812.2 | XP_011525114.1 | 9, 10 | 1568 | |||
XM_011526813.2 | XP_011525115.1 | 9, 10 | 1417 | |||
XM_011526815.2 | XP_011525117.1 | 8, 9 | 2131 | |||
XM_017026646.1 | XP_016882135.1 | 8, 9 | 1992 | |||
GenBank mRNA | AB195682.1 | - | 9, 10 | 1346 | ||
AK301750.1 | - | 8, 9 | 2131 | |||
AY552609.1 | - | 8, 9 | 977 | |||
AY552610.1 | - | 8, 9 | 903 | |||
BC034769.1 | - | 9, 10 | 1488 | 5167853 |