Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199324.1 | NP_001186253.1 | 7 | 730 | ||
NM_001321317.1 | NP_001308246.1 | 4 | 455 | |||
NM_001321318.1 | NP_001308247.1 | 6 | 526 | |||
NM_001321319.1 | NP_001308248.1 | 6 | 692 | |||
NM_001321320.1 | NP_001308249.1 | 5 | 659 | |||
NM_001321321.1 | NP_001308250.1 | 6 | 685 | |||
NM_001321322.1 | NP_001308251.1 | 6 | 680 | |||
NM_001321323.1 | NP_001308252.1 | 5 | 488 | |||
NM_198480.3 | NP_940882.3 | 6 | 697 | |||
XM_011526824.1 | XP_011525126.1 | 8 | 1008 | |||
XM_011526825.1 | XP_011525127.1 | 8 | 1134 | |||
XM_011526826.1 | XP_011525128.1 | 7 | 974 | |||
XM_011526829.1 | XP_011525131.1 | 6 | 743 | |||
XM_011526830.1 | XP_011525132.1 | 5 | 546 | |||
XM_017026648.1 | XP_016882137.1 | 7 | 975 | |||
XM_017026649.1 | XP_016882138.1 | 7 | 1101 | |||
XM_017026650.1 | XP_016882139.1 | 7 | 808 | |||
XM_017026651.1 | XP_016882140.1 | 8 | 1221 | |||
GenBank mRNA | AK131494.1 | - | 6 | 672 | ||
BC053884.1 | - | 6 | 765 | |||
BC105100.1 | - | 4 | 418 | 8144103 | ||
BC105102.1 | - | 4 | 418 | 8144105 | ||
BC143429.1 | - | 5 | 451 | 9051937 | ||
BX647556.1 | - | 6 | 658 |