Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083961.1 | NP_001077430.1 | 10 | 1380 | ||
NM_173636.4 | NP_775907.4 | 10 | 1380 | |||
XM_005258809.2 | XP_005258866.1 | 10 | 1380 | |||
XM_011526837.1 | XP_011525139.1 | 10 | 1365 | |||
XM_011526838.1 | XP_011525140.1 | 10 | 1380 | |||
XM_011526839.1 | XP_011525141.1 | 8 | 1029 | |||
XM_011526840.2 | XP_011525142.1 | 3 | 375 | |||
XM_017026665.1 | XP_016882154.1 | 10 | 1380 | |||
GenBank mRNA | AL041673.1 | - | 4 | 466 | ||
BC058939.1 | - | 10 | 1324 | |||
BX647726.1 | - | 10 | 1299 |