Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145402.1 | NP_001138874.1 | 1 | 320 | ||
GenBank mRNA | BC031875.1 | - | 1 | 309 |