Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321298.1 | NP_001308227.1 | 4, 5 | 708 | ||
NM_173637.3 | NP_775908.2 | 3, 4 | 610 | |||
NR_135612.1 | 3, 4 | 610 | ||||
XM_011527926.1 | XP_011526228.1 | 3, 4 | 404 | |||
GenBank mRNA | AK097761.1 | - | 4, 5 | 692 | ||
BC031671.1 | - | 3, 4 | 610 | 5164630 |