Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NWD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007525.3 | NP_001007526.3 | 14 | 3395 | ||
NM_001290355.1 | NP_001277284.1 | 15 | 4047 | |||
XM_011527929.1 | XP_011526231.1 | 15 | 3579 | |||
XM_011527930.2 | XP_011526232.1 | 15 | 4065 | |||
XM_011527931.2 | XP_011526233.1 | 14 | 3845 | |||
XM_011527934.1 | XP_011526236.1 | 10 | 1784 | |||
XM_011527935.1 | XP_011526237.1 | 15 | 3579 | |||
XM_017026669.1 | XP_016882158.1 | 16 | 4132 | |||
GenBank mRNA | BC117698.1 | - | 13 | 3043 | 40073616 | |
BX648940.1 | - | 16 | 4047 |