Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HKR1 SIRNA Assays ›
Gene Symbol:
HKR1
Gene Name:
HKR1, GLI-Kruppel zinc finger family member
Gene Aliases:
ZNF875
Chromosome Location:
Chr.19: 37317900 - 37364455 on Build GRCh38
UniGene ID:
Hs.657027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s49672
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HKR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_181786.2
NP_861451.1
6
1202
XM_017026672.1
XP_016882161.1
4
1390
XM_017026673.1
XP_016882162.1
4
1456
XM_017026674.1
XP_016882163.1
8
1756
XM_017026675.1
XP_016882164.1
8
1753
XM_017026676.1
XP_016882165.1
5
1051
XM_017026677.1
XP_016882166.1
5
1038
XM_017026678.1
XP_016882167.1
5
1048
XM_017026679.1
XP_016882168.1
5
1035
XM_017026680.1
XP_016882169.1
8
1418
XM_017026681.1
XP_016882170.1
7
1366
XM_017026682.1
XP_016882171.1
8
1415
XM_017026683.1
XP_016882172.1
7
1277
XM_017026684.1
XP_016882173.1
6
1139
XM_017026685.1
XP_016882174.1
4
984
XM_017026686.1
XP_016882175.1
7
1274
XM_017026687.1
XP_016882176.1
6
1136
XM_017026688.1
XP_016882177.1
4
3208
XM_017026689.1
XP_016882178.1
6
1222
XM_017026690.1
XP_016882179.1
4
1002
XM_017026691.1
XP_016882180.1
3
782
XM_017026692.1
XP_016882181.1
8
1352
XM_017026693.1
XP_016882182.1
9
1473
XM_017026694.1
XP_016882183.1
9
1423
XM_017026695.1
XP_016882184.1
8
1395
XM_017026696.1
XP_016882185.1
7
1247
XM_017026697.1
XP_016882186.1
6
1169
XM_017026698.1
XP_016882187.1
6
1147
XM_017026699.1
XP_016882188.1
4
3983
XM_017026700.1
XP_016882189.1
6
1148
XM_017026701.1
XP_016882190.1
8
1346
XM_017026702.1
XP_016882191.1
7
1268
XM_017026703.1
XP_016882192.1
5
1045
XM_017026704.1
XP_016882193.1
5
1107
XM_017026705.1
XP_016882194.1
2
1104
XM_017026707.1
XP_016882196.1
5
1520
XM_017026708.1
XP_016882197.1
5
1523
GenBank mRNA
AB013897.1
-
5
1049
AK056243.1
-
6
1136
AK128116.1
-
9
1755
AK223616.1
-
5
1042
AK299969.1
-
5
1016
AK307852.1
-
1
760
BC004513.2
-
1
224
BC017256.2
-
6
1202
3356680
BC053845.1
-
4
1039
5493685
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
zinc finger transcription factor
C2H2 zinc finger transcription factor
gene-specific transcriptional regulator
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline