Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2L13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304535.1 | NP_001291464.1 | 2 | 949 | ||
NM_175911.3 | NP_787107.1 | 3 | 1074 | |||
XM_011544169.2 | XP_011542471.1 | 3 | 818 | |||
GenBank mRNA | BC028158.1 | - | 3 | 912 | 5240594 |