Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001443.2 | NP_001434.1 | 2 | 275 | ||
GenBank mRNA | BC022287.1 | - | 2 | 260 | ||
BC032801.1 | - | 2 | 186 | 5190267 | ||
BP263155.1 | - | 2 | 119 | |||
CR542090.1 | - | 2 | 161 | |||
M10050.1 | - | 2 | 203 | |||
M10617.1 | - | 2 | 176 |