Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP4A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000778.3 | NP_000769.2 | 11, 12 | 1424 | ||
NM_001319155.1 | NP_001306084.1 | 11, 12 | 1328 | |||
NR_134988.1 | 9, 10 | 1078 | ||||
NR_134989.1 | 10, 11 | 1269 | ||||
NR_134990.1 | 10, 11 | 1263 | ||||
NR_134991.1 | 10, 11 | 1250 | ||||
NR_134992.1 | 7, 8 | 879 | ||||
NR_134993.1 | 8, 9 | 1013 | ||||
NR_134994.1 | 10, 11 | 1285 | ||||
XM_017000464.1 | XP_016855953.1 | 9, 10 | 1122 | |||
XM_017000465.1 | XP_016855954.1 | 11, 12 | 1343 | |||
GenBank mRNA | D26481.1 | - | 11, 12 | 1388 | ||
S67580.1 | - | 11, 12 | 1414 | |||
S67581.1 | - | 11, 12 | 1414 |
Gene Symbol: CYP4A22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010969.3 | NP_001010969.2 | 11, 12 | 1434 | ||
XM_005270767.3 | XP_005270824.1 | 9, 10 | 1122 | |||
GenBank mRNA | AY280371.1 | - | 11, 12 | 1424 | ||
AY280372.1 | - | 11, 12 | 1373 | |||
BC148248.1 | - | 11, 12 | 1434 | 40132876 |