Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161368.1 | NP_001154840.1 | 5 | 458 | ||
NM_174977.3 | NP_777637.1 | 5 | 458 | |||
XM_006724235.2 | XP_006724298.1 | 5 | 474 | |||
XM_011530164.2 | XP_011528466.1 | 5 | 638 | |||
XM_017028787.1 | XP_016884276.1 | 4 | 587 | |||
XM_017028788.1 | XP_016884277.1 | 4 | 588 | |||
XM_017028789.1 | XP_016884278.1 | 4 | 260 | |||
XM_017028790.1 | XP_016884279.1 | 4 | 595 | |||
XM_017028791.1 | XP_016884280.1 | 4 | 287 | |||
GenBank mRNA | AK092002.1 | - | 6 | 507 | ||
AK093418.1 | - | 5 | 475 | |||
AK093448.1 | - | 5 | 442 | |||
AY158085.1 | - | 5 | 358 | |||
BC136358.1 | - | 5 | 430 | 9020345 | ||
BC136359.1 | - | 5 | 430 | 9020346 | ||
BC139912.1 | - | 5 | 397 | 40147686 | ||
CR456346.1 | - | 3 | 217 |