Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080539.1 | NP_001074008.1 | 26 | 3168 | ||
XM_006712438.3 | XP_006712501.1 | 25 | 3105 | |||
XM_006712439.3 | XP_006712502.1 | 10 | 1752 | |||
XM_011510985.2 | XP_011509287.1 | 24 | 2897 | |||
XM_011510987.2 | XP_011509289.1 | 18 | 2088 | |||
XM_011510988.2 | XP_011509290.1 | 22 | 2614 | |||
XM_017003861.1 | XP_016859350.1 | 24 | 3012 | |||
XM_017003862.1 | XP_016859351.1 | 24 | 2991 | |||
XM_017003863.1 | XP_016859352.1 | 24 | 2910 | |||
XM_017003865.1 | XP_016859354.1 | 22 | 2629 | |||
XM_017003866.1 | XP_016859355.1 | 19 | 2304 | |||
GenBank mRNA | AK001064.1 | - | 3 | 1297 | ||
AK125768.1 | - | 10 | 2136 | |||
AK302685.1 | - | 21 | 2536 | |||
AL834537.1 | - | 9 | 2052 |