Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127505.2 | NP_001120977.1 | 6 | 615 | ||
NM_004315.5 | NP_004306.3 | 6 | 681 | |||
NM_177924.4 | NP_808592.2 | 6 | 748 | |||
XM_005273504.3 | XP_005273561.1 | 6 | 509 | |||
GenBank mRNA | AK025211.1 | - | 1 | 764 | ||
AK025732.1 | - | 7 | 651 | |||
AK222780.1 | - | 7 | 484 | |||
AK289481.1 | - | 7 | 649 | |||
AY305384.1 | - | 6 | 522 | |||
BC016481.1 | - | 7 | 471 | 3923451 | ||
BC016828.1 | - | 6 | 602 | 4109317 | ||
BC035453.1 | - | 1 | 46 | |||
BG714023.1 | - | 6 | 495 | |||
U47674.1 | - | 6 | 452 | |||
U70063.1 | - | 7 | 453 |