Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM126B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321618.1 | NP_001308547.1 | 11 | 1299 | ||
NM_001321619.1 | NP_001308548.1 | 12 | 1510 | |||
NM_001321621.1 | NP_001308550.1 | 11 | 1412 | |||
NM_001321622.1 | NP_001308551.1 | 13 | 1628 | |||
NM_001321623.1 | NP_001308552.1 | 13 | 1565 | |||
NM_001321624.1 | NP_001308553.1 | 12 | 1467 | |||
NM_001321625.1 | NP_001308554.1 | 13 | 1678 | |||
NM_001321626.1 | NP_001308555.1 | 13 | 1698 | |||
NM_001321627.1 | NP_001308556.1 | 14 | 1621 | |||
NM_001321628.1 | NP_001308557.1 | 14 | 1734 | |||
NM_001321629.1 | NP_001308558.1 | 13 | 1453 | |||
NM_173822.4 | NP_776183.1 | 12 | 1397 | |||
XM_006712443.3 | XP_006712506.1 | 12 | 1580 | |||
XM_011511002.2 | XP_011509304.1 | 13 | 1595 | |||
XM_017003879.1 | XP_016859368.1 | 14 | 1776 | |||
XM_017003880.1 | XP_016859369.1 | 13 | 2190 | |||
XM_017003881.1 | XP_016859370.1 | 12 | 1458 | |||
XM_017003882.1 | XP_016859371.1 | 14 | 1677 | |||
XM_017003883.1 | XP_016859372.1 | 13 | 1852 | |||
GenBank mRNA | AK097111.1 | - | 5 | 640 | ||
AK315104.1 | - | 12 | 1391 | |||
AL832234.1 | - | 11 | 1256 | |||
AL832346.1 | - | 14 | 1616 | |||
AL833403.1 | - | 12 | 1381 | |||
BC039295.1 | - | 12 | 1395 | 5267063 |