Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033575.1 | NP_001028747.1 | 3 | 1268 | ||
GenBank mRNA | BC036198.1 | - | 2 | 3173 | 5298326 |