Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173653.3 | NP_775924.1 | 2 | 559 | ||
XM_011512704.2 | XP_011511006.1 | 2 | 532 | |||
XM_017006202.1 | XP_016861691.1 | 2 | 532 | |||
GenBank mRNA | AB089794.1 | - | 2 | 350 | ||
AK315107.1 | - | 2 | 486 | |||
AL832304.1 | - | 2 | 424 | |||
AY254100.1 | - | 2 | 474 | |||
BC035779.1 | - | 2 | 475 | 5590356 | ||
DA387367.1 | - | 2 | 532 |