Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080448.2 | NP_001073917.2 | 10 | 2132 | ||
NM_001278300.1 | NP_001265229.1 | 7 | 544 | |||
NM_173655.3 | NP_775926.1 | 8 | 660 | |||
XM_006713592.3 | XP_006713655.1 | 11 | 2180 | |||
XM_017006210.1 | XP_016861699.1 | 11 | 2180 | |||
XM_017006211.1 | XP_016861700.1 | 12 | 2205 | |||
XM_017006212.1 | XP_016861701.1 | 11 | 1874 | |||
XM_017006213.1 | XP_016861702.1 | 11 | 2232 | |||
XM_017006214.1 | XP_016861703.1 | 10 | 1500 | |||
XM_017006215.1 | XP_016861704.1 | 6 | 529 | |||
XM_017006216.1 | XP_016861705.1 | 5 | 479 | |||
XM_017006217.1 | XP_016861706.1 | 7 | 576 | |||
XM_017006218.1 | XP_016861707.1 | 5 | 379 | |||
XM_017006220.1 | XP_016861709.1 | 7 | 544 | |||
XM_017006221.1 | XP_016861710.1 | 5 | 490 | |||
XM_017006222.1 | XP_016861711.1 | 6 | 538 | |||
XM_017006223.1 | XP_016861712.1 | 7 | 544 | |||
GenBank mRNA | AK302040.1 | - | 7 | 527 | ||
AK302080.1 | - | 6 | 434 | |||
AK310249.1 | - | 6 | 463 | |||
AL133666.1 | - | 7 | 607 | |||
DC398430.1 | - | 6 | 482 |