Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C3orf38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173824.3 | NP_776185.2 | 3 | 920 | ||
XM_005264745.4 | XP_005264802.1 | 3 | 946 | |||
GenBank mRNA | AK297719.1 | - | 3 | 570 | ||
AK313547.1 | - | 3 | 661 | |||
AL832398.1 | - | 3 | 920 | |||
BC024188.2 | - | 3 | 704 | 4824439 | ||
KU178900.1 | - | 3 | 603 | |||
KU178901.1 | - | 3 | 510 |