Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256613.1 | NP_001243542.1 | 9 | 1994 | ||
NM_001256614.1 | NP_001243543.1 | 7 | 1800 | |||
NM_182589.2 | NP_872395.2 | 8 | 1767 | |||
NM_198313.2 | NP_938055.1 | 8 | 1949 | |||
NM_198314.2 | NP_938056.1 | 7 | 1722 | |||
GenBank mRNA | DQ644022.1 | - | 9 | 1994 |