Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256613.1 | NP_001243542.1 | 2 | 604 | ||
NM_001256614.1 | NP_001243543.1 | 1 | 377 | |||
NM_182589.2 | NP_872395.2 | 1 | 377 | |||
NM_198313.2 | NP_938055.1 | 2 | 604 | |||
NM_198314.2 | NP_938056.1 | 1 | 377 | |||
GenBank mRNA | AY159813.2 | - | 1 | 377 | ||
AY349352.1 | - | 2 | 138 | |||
AY349353.1 | - | 1 | 183 | |||
BC101185.2 | - | 2 | 156 | 40017964 | ||
DQ644022.1 | - | 2 | 604 | |||
EU165354.1 | - | 1 | 183 |