Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320531.1 | NP_001307460.1 | 24 | 3292 | ||
NM_183061.2 | NP_898884.1 | 25 | 3436 | |||
NR_135297.1 | 19 | 2495 | ||||
XM_006713603.2 | XP_006713666.1 | 25 | 3471 | |||
XM_011512718.1 | XP_011511020.1 | 24 | 3274 | |||
XM_011512719.1 | XP_011511021.1 | 24 | 3265 | |||
XM_011512721.1 | XP_011511023.1 | 26 | 3614 | |||
XM_011512725.2 | XP_011511027.1 | 15 | 1985 | |||
XM_017006246.1 | XP_016861735.1 | 23 | 3139 | |||
XM_017006247.1 | XP_016861736.1 | 24 | 3250 | |||
XM_017006248.1 | XP_016861737.1 | 22 | 3040 | |||
GenBank mRNA | AK128084.1 | - | 25 | 3436 | ||
BC044801.1 | - | 24 | 3261 | 4827529 | ||
BC143774.1 | - | 18 | 2340 |