Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAIM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029839.2 | NP_001025010.1 | 3 | 487 | ||
NM_001029840.2 | NP_001025011.1 | 3 | 305 | |||
NM_001282913.1 | NP_001269842.1 | 3 | 305 | |||
NM_001282914.1 | NP_001269843.1 | 3 | 487 | |||
NM_001282915.1 | NP_001269844.1 | 3 | 305 | |||
NM_173826.3 | NP_776187.2 | 3 | 487 | |||
XM_017006251.1 | XP_016861740.1 | 3 | 532 | |||
GenBank mRNA | AK056593.1 | - | 3 | 487 | ||
AK292756.1 | - | 3 | 305 | |||
AL832473.1 | - | 3 | 164 | |||
BC009400.2 | - | 3 | 283 | |||
BC038420.1 | - | 3 | 407 | |||
BC047231.1 | - | 3 | 182 | 4817904 | ||
BC062587.1 | - | 3 | 459 | |||
BC073829.1 | - | 3 | 132 | |||
BC101058.2 | - | 3 | 164 | |||
BC101059.2 | - | 3 | 164 | 40008143 | ||
BC101060.2 | - | 3 | 164 | 40008144 | ||
BC101061.2 | - | 3 | 164 | 40008152 | ||
CN372173.1 | - | 3 | 260 | |||
DC413255.1 | - | 3 | 305 |