Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198485.3 | NP_940887.1 | 6 | 909 | ||
XM_005247382.4 | XP_005247439.1 | 6 | 4153 | |||
XM_011512732.2 | XP_011511034.2 | 7 | 3039 | |||
XM_011512735.2 | XP_011511037.1 | 5 | 3977 | |||
XM_017006257.1 | XP_016861746.1 | 14 | 2201 | |||
XM_017006258.1 | XP_016861747.1 | 11 | 1629 | |||
XM_017006259.1 | XP_016861748.1 | 13 | 2812 | |||
XM_017006260.1 | XP_016861749.1 | 13 | 2008 | |||
XM_017006261.1 | XP_016861750.1 | 10 | 1757 | |||
XM_017006262.1 | XP_016861751.1 | 7 | 1047 | |||
XM_017006263.1 | XP_016861752.1 | 5 | 625 | |||
GenBank mRNA | AK123232.1 | - | 11 | 1968 | ||
AK125682.1 | - | 6 | 910 | |||
BC101550.1 | - | 6 | 810 | 8069056 | ||
BC101552.1 | - | 6 | 810 | 8069058 |