Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308081.1 | NP_001295010.1 | 6 | 699 | ||
NM_173666.3 | NP_775937.1 | 6 | 920 | |||
XM_011543338.2 | XP_011541640.2 | 7 | 2313 | |||
XM_011543340.2 | XP_011541642.1 | 7 | 812 | |||
GenBank mRNA | AK091296.1 | - | 6 | 860 | ||
BC114508.1 | - | 6 | 856 | 40024023 | ||
BC114955.1 | - | 6 | 856 | 40024022 | ||
BX648369.1 | - | 6 | 699 |
Gene Symbol: SLFN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005257995.4 | XP_005258052.1 | 6 | 4457 | ||
XM_011524967.2 | XP_011523269.1 | 7 | 4579 | |||
XM_017024809.1 | XP_016880298.1 | 5 | 4390 | |||
XM_017024810.1 | XP_016880299.1 | 6 | 5047 |