Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004457.3 | NP_004448.2 | 11 | 1662 | ||
NM_203372.1 | NP_976251.1 | 10 | 1555 | |||
XM_017003584.1 | XP_016859073.1 | 9 | 1474 | |||
GenBank mRNA | AB061712.1 | - | 11 | 1630 | ||
AK001471.1 | - | 6 | 581 | |||
AK023191.1 | - | 8 | 1207 | |||
AK314621.1 | - | 11 | 1640 | |||
BC041692.1 | - | 11 | 1654 | 5226905 | ||
D89053.1 | - | 9 | 1353 |