Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC36A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145017.1 | NP_001138489.1 | 11 | 1875 | ||
NM_181774.3 | NP_861439.3 | 10 | 1752 | |||
XM_011537627.2 | XP_011535929.1 | 10 | 1674 | |||
XM_011537629.2 | XP_011535931.1 | 9 | 1582 | |||
XM_011537630.1 | XP_011535932.1 | 8 | 1099 | |||
XM_011537631.1 | XP_011535933.1 | 8 | 1110 | |||
XM_011537633.1 | XP_011535935.1 | 6 | 962 | |||
GenBank mRNA | AK127978.1 | - | 10 | 1745 | ||
AK131483.1 | - | 9 | 3233 | |||
AY162215.1 | - | 10 | 1570 | |||
AY228365.1 | - | 3 | 379 | |||
BC101092.1 | - | 11 | 1598 | 40009824 | ||
BC101093.1 | - | 11 | 1598 | 40009826 | ||
BC101094.1 | - | 10 | 1475 | 40009832 | ||
BC101095.2 | - | 10 | 1475 | 40009834 |