Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099293.1 | NP_001092763.1 | 1 | 1797 | ||
GenBank mRNA | AK302388.1 | - | 1 | 1742 |