Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000527.4 | NP_000518.1 | 4 | 864 | ||
NM_001195798.1 | NP_001182727.1 | 4 | 864 | |||
NM_001195799.1 | NP_001182728.1 | 3 | 741 | |||
XM_011528010.1 | XP_011526312.1 | 4 | 827 | |||
GenBank mRNA | AB209409.1 | - | 4 | 748 | ||
AK295612.1 | - | 3 | 400 | |||
AK300313.1 | - | 3 | 640 | |||
AY114155.1 | - | 4 | 691 | |||
BC014514.1 | - | 4 | 747 | 3542845 | ||
BT007361.1 | - | 4 | 677 | |||
BX648281.1 | - | 4 | 932 |