Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002014.3 | NP_002005.1 | 5 | 852 | ||
XM_011520929.2 | XP_011519231.1 | 5 | 1845 | |||
GenBank mRNA | AK313918.1 | - | 5 | 817 | ||
BC001786.1 | - | 5 | 807 | 3542330 | ||
BC002887.2 | - | 2 | 211 | |||
BC007924.2 | - | 5 | 752 | 4304462 | ||
M88279.1 | - | 5 | 753 |