Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173829.3 | NP_776190.1 | 3 | 310 | ||
XM_011543344.2 | XP_011541646.1 | 2 | 442 | |||
GenBank mRNA | AK094073.1 | - | 310 | |||
AL832348.1 | - | 3 | 267 | |||
BC047006.1 | - | 3 | 239 | |||
BC108716.1 | - | 3 | 240 | 6184559 | ||
BC127085.1 | - | 3 | 248 | 40122236 | ||
BC127086.1 | - | 3 | 248 | 40122239 |