Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCBLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173674.2 | NP_775945.1 | 5 | 746 | ||
XM_005266941.1 | XP_005266998.1 | 5 | 710 | |||
XM_006715463.1 | XP_006715526.1 | 5 | 710 | |||
XM_011535771.1 | XP_011534073.1 | 6 | 795 | |||
XM_011535773.2 | XP_011534075.1 | 5 | 710 | |||
XM_011535774.2 | XP_011534076.1 | 5 | 710 | |||
XM_017010775.1 | XP_016866264.1 | 6 | 719 | |||
GenBank mRNA | AK095973.1 | - | 5 | 616 | ||
BC035671.1 | - | 5 | 693 | 5730536 |