Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREML4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198153.2 | NP_937796.1 | 2 | 414 | ||
GenBank mRNA | AF534826.1 | - | 1 | 214 | ||
AY358271.1 | - | 2 | 417 | |||
BC101780.1 | - | 2 | 402 | 8069286 | ||
BC105101.1 | - | 2 | 402 | 8144104 | ||
BC143652.1 | - | 2 | 402 | 9052166 |