Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCAF2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110549.1 | 3, 4 | 1302 | |||
GenBank mRNA | BG748361.1 | - | 3, 4 | 645 | ||
CB046449.1 | - | 2, 3 | 320 |
Gene Symbol: TCAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130025.1 | NP_001123497.1 | 5, 6 | 2008 | ||
NM_001130026.2 | NP_001123498.2 | 5, 6 | 1637 | |||
NM_173678.2 | NP_775949.2 | 5, 6 | 1994 | |||
XM_006715927.3 | XP_006715990.1 | 4, 5 | 2869 | |||
XM_006715928.3 | XP_006715991.1 | 4, 5 | 2869 | |||
XM_011516059.2 | XP_011514361.1 | 4, 5 | 1457 | |||
GenBank mRNA | AK090395.1 | - | 3, 4 | 1100 | ||
AK098041.1 | - | 4, 5 | 1368 | |||
AK296318.1 | - | 5, 6 | 1613 | |||
AL832734.1 | - | 5, 6 | 1989 | |||
AY167570.1 | - | 5, 6 | 1991 | |||
BC113530.1 | - | 5, 6 | 1994 | 8322582 | ||
BC117233.1 | - | 5, 6 | 1994 | 40125784 |