Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NSMCE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173685.2 | NP_775956.1 | 4 | 476 | ||
XM_005250875.2 | XP_005250932.1 | 4 | 484 | |||
XM_005250876.4 | XP_005250933.1 | 3 | 377 | |||
XM_011516974.2 | XP_011515276.1 | 5 | 546 | |||
XM_011516975.1 | XP_011515277.1 | 4 | 441 | |||
XM_017013330.1 | XP_016868819.1 | 2 | 657 | |||
XM_017013331.1 | XP_016868820.1 | 5 | 514 | |||
XM_017013332.1 | XP_016868821.1 | 2 | 675 | |||
XM_017013333.1 | XP_016868822.1 | 5 | 586 | |||
XM_017013334.1 | XP_016868823.1 | 2 | 697 | |||
GenBank mRNA | AK057002.1 | - | 4 | 446 | ||
BC032797.1 | - | 4 | 475 | 5109691 |