Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF707 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100598.1 | NP_001094068.1 | 6 | 549 | ||
NM_001100599.1 | NP_001094069.1 | 5 | 451 | |||
NM_001288805.1 | NP_001275734.1 | 8 | 1263 | |||
NM_001288806.1 | NP_001275735.1 | 6 | 549 | |||
NM_001288807.1 | NP_001275736.1 | 6 | 537 | |||
NM_001288808.1 | NP_001275737.1 | 4 | 384 | |||
NM_001288809.1 | NP_001275738.1 | 4 | 558 | |||
NM_173831.3 | NP_776192.2 | 7 | 623 | |||
NR_110190.1 | 5 | 484 | ||||
NR_110191.1 | 6 | 582 | ||||
NR_110192.1 | 5 | 487 | ||||
NR_110193.1 | 6 | 582 | ||||
XM_005250883.3 | XP_005250940.1 | 6 | 570 | |||
XM_006716542.3 | XP_006716605.1 | 7 | 639 | |||
XM_006716543.3 | XP_006716606.1 | 6 | 544 | |||
XM_011516977.2 | XP_011515279.1 | 6 | 728 | |||
XM_011516979.2 | XP_011515281.1 | 7 | 589 | |||
XM_017013335.1 | XP_016868824.1 | 8 | 746 | |||
GenBank mRNA | AK001126.1 | - | 5 | 480 | ||
AK055198.1 | - | 3 | 1369 | |||
AK055263.1 | - | 5 | 525 | |||
AK095138.1 | - | 8 | 1207 | |||
AK290432.1 | - | 6 | 520 | |||
BC014894.1 | - | 7 | 583 | 3910185 | ||
CV023324.1 | - | 3 | 302 | |||
DA007942.1 | - | 6 | 520 | |||
DA188478.1 | - | 5 | 487 | |||
DA917297.1 | - | 5 | 399 | |||
DA957023.1 | - | 6 | 510 | |||
DA990503.1 | - | 6 | 494 | |||
DB099015.1 | - | 6 | 555 | |||
DB106612.1 | - | 4 | 328 |