Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MICU3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181723.2 | NP_859074.1 | 11 | 1225 | ||
XM_005273477.2 | XP_005273534.1 | 10 | 1184 | |||
XM_006716328.2 | XP_006716391.1 | 10 | 1127 | |||
XM_006716329.2 | XP_006716392.1 | 9 | 1088 | |||
XM_011544507.2 | XP_011542809.1 | 11 | 1205 | |||
XM_017013336.1 | XP_016868825.1 | 10 | 1166 | |||
XM_017013340.1 | XP_016868829.1 | 10 | 1170 | |||
XM_017013342.1 | XP_016868831.1 | 9 | 1037 | |||
GenBank mRNA | AL833419.1 | - | 1 | 109 | ||
BC032868.1 | - | 11 | 1180 | |||
BC045553.1 | - | 11 | 1224 | 5262603 |