Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 3, 4 | 583 | ||
NR_126399.1 | 2, 3 | 438 | ||||
XM_017013346.1 | XP_016868835.1 | 2, 3 | 189 | |||
GenBank mRNA | AK094450.1 | - | 3, 4 | 531 | ||
BC063497.1 | - | 2, 3 | 349 | |||
BC120993.2 | - | 2, 3 | 373 | |||
DB260703.1 | - | 2, 3 | 438 |