Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000086.2 | NP_000077.1 | 4 | 581 | ||
NM_001042432.1 | NP_001035897.1 | 5 | 617 | |||
NM_001286105.1 | NP_001273034.1 | 3 | 502 | |||
NM_001286110.1 | NP_001273039.1 | 3 | 502 | |||
GenBank mRNA | AF015593.1 | - | 4 | 312 | ||
AF015594.1 | - | 3 | 273 | |||
AF015595.1 | - | 3 | 276 | |||
AF015597.1 | - | 4 | 331 | |||
AF015598.1 | - | 4 | 363 | |||
AF015600.1 | - | 3 | 285 | |||
AF015602.1 | - | 2 | 149 | |||
AF077956.1 | - | 4 | 258 | |||
AF077957.1 | - | 4 | 258 | |||
AF077958.1 | - | 4 | 258 | |||
AF077959.1 | - | 4 | 258 | |||
AF077960.1 | - | 4 | 258 | |||
AF077961.1 | - | 4 | 258 | |||
AF077962.1 | - | 4 | 258 | |||
AF077963.1 | - | 4 | 258 | |||
AF077964.1 | - | 4 | 258 | |||
AF077965.1 | - | 4 | 258 | |||
AF077966.1 | - | 4 | 258 | |||
AF077967.1 | - | 4 | 258 | |||
AF077968.1 | - | 4 | 258 | |||
AF077971.1 | - | 4 | 258 | |||
AF077972.1 | - | 4 | 258 | |||
AF077973.1 | - | 3 | 179 | |||
AF078169.1 | - | 4 | 258 | |||
AK090709.1 | - | 6 | 1612 | |||
AK294070.1 | - | 3 | 426 | |||
AK294250.1 | - | 5 | 387 | |||
AK297690.1 | - | 3 | 444 | |||
AK302138.1 | - | 4 | 306 | |||
AK313002.1 | - | 5 | 389 | |||
BC002394.2 | - | 5 | 375 | 2820893 | ||
BC004433.1 | - | 5 | 375 | 2820893 | ||
BP252176.1 | - | 5 | 397 | |||
U32680.1 | - | 4 | 395 |