Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CLN3 SIRNA Assays ›
Gene Symbol:
CLN3
Gene Name:
CLN3, battenin
Gene Aliases:
BTS, JNCL
Chromosome Location:
Chr.16: 28466653 - 28492302 on Build GRCh38
UniGene ID:
Hs.534667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s501293
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CLN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000086.2
NP_000077.1
11
1191
NM_001042432.1
NP_001035897.1
12
1227
NM_001286104.1
NP_001273033.1
11
1155
NM_001286105.1
NP_001273034.1
9
1032
NM_001286109.1
NP_001273038.1
9
1040
NM_001286110.1
NP_001273039.1
10
1112
GenBank mRNA
AF015593.1
-
11
922
AF015594.1
-
10
883
AF015595.1
-
9
742
AF015597.1
-
9
724
AF015598.1
-
9
676
AF015600.1
-
7
598
AF015601.1
-
6
512
AF077956.1
-
11
868
AF077957.1
-
11
868
AF077958.1
-
11
868
AF077959.1
-
11
868
AF077960.1
-
10
724
AF077961.1
-
11
868
AF077962.1
-
11
868
AF077963.1
-
11
868
AF077964.1
-
9
651
AF077965.1
-
8
538
AF077966.1
-
11
868
AF077967.1
-
10
788
AF077968.1
-
9
651
AF077969.1
-
7
482
AF077970.1
-
7
482
AF077971.1
-
11
868
AF077973.1
-
9
709
AF078169.1
-
11
868
AK090709.1
-
12
2302
AK294070.1
-
10
1036
AK294250.1
-
10
997
AK297690.1
-
9
974
AK302027.1
-
11
895
AK302138.1
-
11
916
AK313002.1
-
12
999
BC002394.2
-
12
985
2820893
BC004433.1
-
12
985
2820893
BC111068.1
-
9
970
5764535
EF587244.1
-
4
245
U32680.1
-
11
1005
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
action potential
globoside metabolic process
cellular amino acid metabolic process
ceramide metabolic process
glucosylceramide metabolic process
galactosylceramide metabolic process
sphingomyelin metabolic process
receptor-mediated endocytosis
vacuolar transport
lysosome organization
lysosomal lumen acidification
associative learning
arginine transport
macroautophagy
negative regulation of macroautophagy
protein processing
protein catabolic process
ionotropic glutamate receptor signaling pathway
lysosomal lumen pH elevation
neurotransmitter metabolic process
amyloid precursor protein catabolic process
negative regulation of apoptotic process
negative regulation of catalytic activity
negative regulation of neuron apoptotic process
negative regulation of proteolysis
vesicle transport along microtubule
neuromuscular process controlling balance
regulation of cytosolic calcium ion concentration
membrane organization
autophagosome maturation
protein binding
unfolded protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0