Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP27A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000784.3 | NP_000775.1 | 4 | 1167 | ||
XM_017003488.1 | XP_016858977.1 | 3 | 1719 | |||
GenBank mRNA | AK226039.1 | - | 4 | 1168 | ||
AK290418.1 | - | 4 | 773 | |||
AK310782.1 | - | 2 | 679 | |||
AY178622.1 | - | 4 | 733 | |||
BC040430.1 | - | 4 | 747 | 5760376 | ||
BC051851.1 | - | 4 | 745 | 5240515 | ||
M62401.1 | - | 4 | 754 | |||
X59812.1 | - | 4 | 930 |