Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000181.3 | NP_000172.2 | 11 | 1803 | ||
NM_001284290.1 | NP_001271219.1 | 9 | 1365 | |||
NM_001293104.1 | NP_001280033.1 | 10 | 1618 | |||
NM_001293105.1 | NP_001280034.1 | 9 | 1475 | |||
NR_120531.1 | 10 | 1718 | ||||
XM_005250297.3 | XP_005250354.1 | 10 | 1645 | |||
XM_011516113.1 | XP_011514415.1 | 10 | 1547 | |||
XM_011516114.1 | XP_011514416.1 | 10 | 1475 | |||
XM_017012091.1 | XP_016867580.1 | 9 | 1394 | |||
XM_017012092.1 | XP_016867581.1 | 9 | 1381 | |||
XM_017012093.1 | XP_016867582.1 | 9 | 1322 | |||
GenBank mRNA | AK096764.1 | - | 3 | 2061 | ||
AK223406.1 | - | 11 | 1702 | |||
AK300223.1 | - | 11 | 1592 | |||
AK303819.1 | - | 9 | 1262 | |||
AK312752.1 | - | 11 | 1719 | |||
BC014142.2 | - | 11 | 1673 | 4662011 | ||
BM719712.1 | - | 2 | 56 | |||
CD014092.1 | - | 10 | 1500 | |||
CD014093.1 | - | 9 | 1358 | |||
CD014094.1 | - | 10 | 1600 | |||
M10618.1 | - | 2 | 115 | |||
M15182.1 | - | 11 | 1698 |